Scott Safety™ Envoy Radiocom™
Ideal communications system for first responders builds on the features of the E-Z Radiocom
Manufacturer: Scott Health & Safety 805262-09
 View more versions of this product
Catalog No. 19-818-687
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Motorola™ saber radio and handheld portable radios with remote accessory connectors
    Product Type Communications System
    For Use With (Application) Reduces garbled or lost transmission, which translates into greater safety
    Includes 3 modes ems lapel microphone/amplifier, hazmat remote ptt (push-to-talk), adjustable boom speaker and fire voice amplification both with interfaces
    Mounting Type 0.25 turn universal mounting bracket
    Certifications NIOSH, NFPA 1981, ETL, ANSI UL 913
    Alarm Types LED
    Communications system suits volunteer firefighters, EMS personnel, Haz Mat responders, and industrial users. Many more models are available for different radio systems; please contact your Fisher Safety Representative for more information.
    ·    Features a facepiece-mounted microphone, ear speaker boom and a chest-mounted Push-to-Talk (PTT) console
    ·    Chest-mounted PTT console may be used independently as a lapel microphone in nontactical applications
  • Description & Specifications
Related Products
Product Content Feedback