Learn More
Scott Safety™ Liberty™ 2 Mobile Air System
Charges self-contained breathing apparatus
Supplier:  Scott Safety™ AL20251213112
Catalog No. 19-827-304
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Mobile cylinder charge station features compressor mounted on a dual axle trailer
  • Dual powered by electric motor or a Diesel water-cooled engine
  • Light tower generator
  • SCBA storage rack
  • Meets or exceed the requirements at Grade ”E” breathing air per Compressed Gas Association, Commodity Specification, G-7.1
  • Meets all current NFPA, OSHA, NIOSH, State Industrial Safety and Vehicle Code requirements
Specifications
Specifications
AL20251213112