Learn More
Scott Safety™ M120 CBRN Facepiece
Offers a wide range of applications from chemical, biological, radiological and nuclear (CBRN) response to riot control to HAZMAT response
Manufacturer:  Scott Health & Safety 013013
 View more versions of this product
Catalog No. 19-150-1145
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
40mm canister
Wide range of applications from CBRN response to riot control to hazmat response
Medium/Large
1.01 lb.
EPDM
M-120 Facepiece
Only mask
0.46kg
NIOSH-APR and CBRN-APR
Polycarbonate
Accessories