Scott Safety™ P100 High Efficiency Canister
Manufacturer: Scott Health & Safety 805036-02
 View more versions of this product
Catalog No. 19-121-175
  Check Availability
Add to Cart
    Specifications
    Quantity 10 Pack
    For Use With (Equipment) Av-2000™ and av-3000™, scott-o-vista™ and weld-o-vista™ full facepiece with v-bar adapter,Av-2000™ and av-3000™, scott-o-vista™ and weld-o-vista™ full facepiece with v-bar adapter
    Product Type P100 Chin Canister
    For Use With (Application) Ideal for escape/egress from immediately dangerous to life or health (IDLH) environments
    Certifications NIOSH
  • Description & Specifications
Related Products
Product Content Feedback