Learn More
Scott Safety™ ProMask 25 Full Facepiece General Use Respirator
Uniquely contoured sealing edge provides excellent fit
Manufacturer:  Scott Safety™ 013028
 View more versions of this product
Catalog No. 19-120-2862
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
For use with a full range of 742 Twin Cartridges, NIOSH 42 CFR Part 84 approved filters, chemical cartridges for gases/vapors and combination particulate/gas/vapor cartridges.
  • Simple disinfection, decontamination and maintenance
  • Includes speech diaphragm for optimal personal communication
  • Head harness does not interfere with head or ear protection
  • Sweatport in chin pocket provides comfort during extended wear times
  • Easy breathing twin cartridges are swept back out of the line of vision
  • Balanced, lightweight, and ergonomically designed with low profile
  • Wide lens for extended field of vision, available in polycarbonate or hard-coated polycarbonate for scratch resistance
Specifications
Specifications
NIOSH
Full range of 742 twin cartridge
Speech diaphragm
Halo-butyl Blended Elastomer
Headnet
0.005kg
Simple disinfection, decontamination and maintenance
Polyester
Polycarbonate
Small
1.2 lb.
Promask 25