Learn More
Scott Safety™ ProMask 25 Full Facepiece General Use Respirator
Uniquely contoured sealing edge provides excellent fit
Specifications
Certifications/Compliance NIOSH
Product Type Promask 25
For Use With (Application) Simple disinfection, decontamination and maintenance
For Use With (Equipment) Full range of 742 twin cartridge
Harness Style Polyester
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-120-2861
View Documents
Scott Safety™
013023
Medium/Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-120-2862
View Documents
Scott Safety™
013028
Small This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
For use with a full range of 742 Twin Cartridges, NIOSH 42 CFR Part 84 approved filters, chemical cartridges for gases/vapors and combination particulate/gas/vapor cartridges.
  • Simple disinfection, decontamination and maintenance
  • Includes speech diaphragm for optimal personal communication
  • Head harness does not interfere with head or ear protection
  • Sweatport in chin pocket provides comfort during extended wear times
  • Easy breathing twin cartridges are swept back out of the line of vision
  • Balanced, lightweight, and ergonomically designed with low profile
  • Wide lens for extended field of vision, available in polycarbonate or hard-coated polycarbonate for scratch resistance
Specifications
Specifications
NIOSH
Simple disinfection, decontamination and maintenance
Polyester
Halo-butyl Blended Elastomer
Headnet
0.005 kg
Promask 25
Full range of 742 twin cartridge
Speech diaphragm
Polycarbonate
1.2 lb.