Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ Protege ZM Single-Gas Monitors

Easy-to-use, zero-maintenance gas detection solution that delivers high performance

Manufacturer:  Scott Health & Safety 096345902

 View more versions of this product

Catalog No. 19-122-039


  Check Availability
Add to cart

Requires : PC running the Microsoft™ Windows™ operating system and Microsoft .net framework, using Microsoft Windows Xp, Microsoft Windows Vista™, Microsoft Windows 7 and Microsoft Windows 8 programs.

Specifications

Description

  • Small, lightweight (less than 3 oz.) and ergonomically designed
  • Available in oxygen, carbon monoxide and hydrogen sulfide single gas models
  • Monitors have two years of battery and sensor life once activated; H2S and CO monitors include hibernate feature which extends battery life up to one additional year
  • No need to charge or calibrate once activated, can be deployed in field 24 hours a day, every day for two years without maintenance
  • Easy-to-read LCD with multiple display options including real-time gas readings, monitor life remaining or both
  • Three point alarms (high-visibility LED, audible and tactile) with adjustable settings
  • One-button activation, self test at start up and IP66/67 rated
  • Impact-absorbant rubber over mold case with strong, stainless-steel alligator clip to conveniently place monitor anywhere you want
  • Test station to bump, calibrate or hibernate four monitors simultaneously
  • Can be programmed by IR connect communication module

Customizable Settings include:

  • Custom or factory programmed alarm set points
  • User configurable bump and calibration reminders
  • Data-logging capabilities
  • Name or assign monitors using a six-character user ID

Applications:

Ideal for industrial workers and first responders.

Order Info

Protege ZM monitor is supported by a portfolio of accessories including a four-bay test station and IR Connect programmer.

Certifications

CE marked

Compliance

ETL/cETL certification to UL 913 7th, CSA C22.2 No. 157 for use in Class I Division 1 and 2, Groups A, B, C and D hazardous areas; IECEx Ex ia IIC T4 Ga; ATEX Directive (94/9/EC)

Description & Specifications

Specifications

Hydrogen Sulfide (H2S)
Visual (red LED), Audible (95db) and Vibrating
Lithium ion
1.3 in.
LCD
9.4cm
IP 66/67
Hydrogen Sulfide
76.5g
5.6cm
-40° to +50°C
Two years (Hibernate Mode extends life up to one additional year)
CE
3.3cm
3.7 in.
5 to 95% RH (non-condensing)
1 to 100ppm
2.7 oz.
2.2 in.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit