Learn More
Scott Safety™ RevolveAir™ Fill Stations
Economical charge station designed with safety in mind
Supplier: Scott Safety™ AF21102110401
View more versions of this product
Safety Services Available
Overview
Repair/Recertification
Description
Total containment for two cylinders.- With turntable chamber door that allows for simultaneous charging of two breathing air cylinders while a second set of cylinders is being mounted
- Ergonomically designed
- Third party test and certified (NFPA 1901)
- Meet NFPA and NIOSH Standards
- With safety pressure gauges (storage-compressor and cylinder)
- Auxiliary outlet unregulated back, regulated front
- Automatic storage refill valve
Specifications
Stationary, manual filling | |
Four cylinders: 2 filling, 2 for exchange | |
Fill Station |
Breathing Air Cylinders for use with SCBAs | |
AF21102110401 |