Learn More
Scott Safety™ RevolveAir™ Fill Stations
Economical charge station designed with safety in mind
Supplier:  Scott Safety™ AF2110211A401
 View more versions of this product
Catalog No. 19-827-310
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Total containment for two cylinders.
  • With turntable chamber door that allows for simultaneous charging of two breathing air cylinders while a second set of cylinders is being mounted
  • Ergonomically designed
  • Third party test and certified (NFPA 1901) 
  • Meet NFPA and NIOSH Standards
  • With safety pressure gauges (storage-compressor and cylinder)
  • Auxiliary outlet unregulated back, regulated front
  • Automatic storage refill valve
Specifications
Specifications
Stationary, automatic filling
Four cylinders: 2 filling, 2 for exchange
Fill Station
Breathing Air Cylinders for use with SCBAs
AF2110211A401