Learn More
Scott Safety™ RevolveAir™ Fill Stations
Economical charge station designed with safety in mind
Specifications
For Use With (Equipment) | Breathing Air Cylinders for use with SCBAs |
---|---|
Holds | Four cylinders: 2 filling, 2 for exchange |
Product Type | Fill Station |
Catalog Number | Mfr. No. | Connection Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Connection Type | Price | Quantity & Availability | |||||
19-827-307
|
Scott Safety™
AF21102110401 |
Stationary, manual filling | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
19-827-310
|
Scott Safety™
AF2110211A401 |
Stationary, automatic filling | Item Discontinued This item has been discontinued by the manufacturer and is no longer available. Please reach out to your sales representative. | N/A | |||||
Description
Total containment for two cylinders.- With turntable chamber door that allows for simultaneous charging of two breathing air cylinders while a second set of cylinders is being mounted
- Ergonomically designed
- Third party test and certified (NFPA 1901)
- Meet NFPA and NIOSH Standards
- With safety pressure gauges (storage-compressor and cylinder)
- Auxiliary outlet unregulated back, regulated front
- Automatic storage refill valve
Specifications
Breathing Air Cylinders for use with SCBAs | |
Four cylinders: 2 filling, 2 for exchange | |
Fill Station |