Scott Safety™ Ska-Pak™ 5- and 10-Minute Type C Emergency Escape Breathing Respirators: Facepieces

Manufacturer: Scott Health & Safety 80433031

 View all options for this product

Catalog No. 19-087702

  Check Availability
Add to Cart


    Diameter (English) 3.8 in.
    Diameter (Metric) 10cm
    Cycle Time 10 min.
    Product Type Facepiece
    Flow Rate 200L/min.
    For Use With (Application) Personnel engaged in industrial, hazmat and firefighting operation and hazardous atmospheres, including those identified as immediately dangerous to life or health
    Includes Scott-O-Vista Facepiece; With Hansen fittings
    Weight (English) 9.8 lb.
    Weight (Metric) 4.5kg
    Certifications NIOSH MSHA

    Offer emergency respirator protection for personnel engaged in industrial, Haz Mat, and firefighting operations. Ideal for use in hazardous atmospheres, including those identified as Immediately Dangerous to Life or Health (IDLH)—i.e., environments with little or no oxygen, with high concentrations of contaminants, or where concentrations of contaminants are unknown.

    Choice of AV-2000™ compact, lightweight facepiece with dual voicemitters or Scott-O-Vista™ full-facepiece with cone-shaped lens and net style head harness. Ska-Pak 10-minute Entry/Egress SCBA. Ideal for confined spaces, emergency escape from large industrial plants, Haz Mat remediation applications, and firefighting involving aerial ladders with an air supply line. Unit features a 4500psi high pressure air cylinder that provides 10 minutes of breathable air for entering or exiting hazardous environments. Slim cylinder is constructed of lightweight aluminum with fiberglass overwrap for greater comfort, less fatigue, and personal endurance in labor-intense environments. Aluminum construction enhances service life, eliminating the possibility of internal rust or loose scale. Cylinder is mounted securely on SCBA harness and worn on the hip. Lightweight and low profile design allows easy movement. L x dia. of cylinder: 181/4 x 37/8 in. (46 x 10cm). Ska-Pak harness is constructed of your choice of either Kevlar or polypropylene webbing and consists of an adjustable shoulder strap and waist belt. Both are equipped with corrosion-resistant web adjusters and quick-release buckles for a secure, comfortable fit and easy donning and doffing. Comes with E-Z Flo™ regulator to control airflow in excess of 200L/minute and airline connection with your choice of Hansen or Schrader fittings. Available with your choice of AV-2000 or Scott-O-Vista facepiece. System weight with AV-2000 facepiece: 101/8 lb. (4.6kg); with Scott-O-Vista facepiece: 97/8 lb. (4.5kg). NIOSH/MSHA approved.Ska-Pak 5-minute Entry/Egress SCBA. Uses a lightweight, 2216psi low- pressure cylinder that provides five minutes of breathable air for escape purposes and can be connected to an external air supply for entering and working in hazardous areas. All 5-minute Ska-Pak SCBAs come withE-Z Flo regulator, airline connection with your choice of Hansen or Schrader fittings, and your choice of either the AV-2000 or Scott-O-Vista facepiece. System weight with AV-2000 facepiece: 85/8 lb. (3.9kg); with Scott-O-Vista facepiece: 83/8 lb. (3.8kg). L x dia. of cylinder: 123/8 x 43/8 in. (31 x 11cm). NIOSH/MSHA approved.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit