Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ Ska-Pak™ AT Escape SCBA/Supplied-Air Respirator

Supplied-air respirator offers automatic, hands-free transfer capability for panic-proof protection

$2931.00

Specifications

Certifications/Compliance NIOSH, NFPA
Product Type SCBA/Supplied Air Respirator
For Use With (Application) Use in hazardous industrial, confined space and IDLH environments
Pressure 80 to 115 psi
View More Specs

Products 1
Catalog Number Mfr. No. Cycle Time Includes Price Quantity & Availability  
Catalog Number Mfr. No. Cycle Time Includes Price Quantity & Availability  
19-121-775
View Documents
Scott Safety™
SAR423030431101
10 min. 3000 psi composite cylinder, hansen fitting, av-2000™ facepiece with kevlar headnet, e-z flo™ regulator
Each for $2,931.00
Add to cart
 
Description

Description

Type C airline respirator instantly switches from external air to cylinder air if continuous air supply is interrupted. A Vibralert™ tactile alarm warns the user when transfer occurs; a flashing electronic indicator notifies wearer of low cylinder pressure. Suitable for numerous applications in hazardous industrial, confined space, and IDLH environments.

Components

  • Universal-sized AV-2000™ facepiece with Kevlar™ head harness E-Z Flo™ regulator 5-, 10- and 15-minute portable escape cylinder (3000psig composite) E-Z Klip™ cylinder retention system for quick, easy cylinder changes Choice of nylon or Kevlar harness
Specifications
    Operates at an airline inlet pressure of 80 to 115psig Transfer to cylinder occurs when pressure drops below 50psig Great for aerial apparatus

Certifications

NIOSH approved.

Compliance

Meets NFPA airflow requirements.

Specifications

Specifications

NIOSH, NFPA
SCBA/Supplied Air Respirator
Use in hazardous industrial, confined space and IDLH environments
80 to 115 psi
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit