Scott™ Safety SKA-PAK™ Automatic Transfer Supplied Air Respirator
Manufacturer: Scott Health & Safety 200063-50
 View more versions of this product
Catalog No. 19-121-772
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Confined space applications
    Includes Kevlar harness, 2216psi aluminum bottle, av-2000™ facepiece with kevlar head harness, comfort seal, standard regulator and hansen fitting, vibralert™ tactile alarm system
    Certifications NIOSH
    Product Type Automatic Transfer Supplied Air Respirator
    Duration 3 min.
  • Description & Specifications
Related Products
Product Content Feedback