Learn More
Scott Safety™ Speed-Evac™ Mouthbit Respirator
Designed for emergency escape respiratory protection against a variety of toxic gases in low level concentrations
Supplier: Scott Safety™ 80383430
Description
- Small, lightweight and ergonmically designed so it can be donned for immediate emergency escape protection
- One size fits all
- Requires minimal training and is completely disposable
- comfortable mouthbit, chin rest ane easy-to-use nose clip
- Choice of disposable plastic pouch or reusable ABS case
Specifications
Mouth-Bit Respirator | |
Disposable pouch | |
Universal |
Acid gas, immediate emergency escape protection | |
30/Pk. |