Learn More
Scott Safety™ Speed-Evac™ Mouthbit Respirator
Designed for emergency escape respiratory protection against a variety of toxic gases in low level concentrations
Supplier:  Scott Safety™ 80383403
 View more versions of this product
Catalog No. 19-818-563
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Small, lightweight and ergonmically designed so it can be donned for immediate emergency escape protection
  • One size fits all
  • Requires minimal training and is completely disposable
  • comfortable mouthbit, chin rest ane easy-to-use nose clip
  • Choice of disposable plastic pouch or reusable ABS case
Specifications
Specifications
Mouth-Bit Respirator
Reusable hard case
Universal
Acid gas, non IDLH environments
1 Ea.