Learn More
Scott Safety™ Speed-Evac™ Mouthbit Respirator
Designed for emergency escape respiratory protection against a variety of toxic gases in low level concentrations
Manufacturer:  Scott Safety™ 80383901
 View more versions of this product
Catalog No. 19-818-564
Add to cart
Description
Description
  • Small, lightweight and ergonmically designed so it can be donned for immediate emergency escape protection
  • One size fitss all
  • Requires minimal training and is completely disposable
  • comfortable mouthbit, chin rest ane easy-to-use nose clip
  • Choice of disposable plastic pouch or reusable ABS case
Specifications
Specifications
NIOSH
Mouth-Bit Escape Respirator
Ammonia, non IDLH environments
Universal
Each
Yes
Disposable pouch