Learn More
Scott Safety™ Swat-Pak Accessories, Carrying Case
Carrying and transporting case for SCBA.
Supplier: Scott Safety™ 7012621284
Description
- Carrying case ideal for transporting and storing
- For use with SCBA AP50, NXG, & AP75 models
- Hard molded case made of HDPE and has two-piece replaceable latches
- Heavy-duty soft case made of nylon with zipper
Specifications
SCBA Carry Case | |
80449701 | |
SCBA Air-Pak™ 50 and 75 models, NXG |
HDPE | |
Two-piece Hinge and Replaceable Latches |