Learn More
Scott Safety™ Swat-Pak™ SCBA First Responder Edition
Designed for optimum protection protection during high risk search and rescue operations, hazardous chemicals or biological environments
Manufacturer:  Scott Safety™ 805454416
 View more versions of this product
Catalog No. 19-121-444
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Swat-Pak SCBA, AV-2000 Comfort Seal facepiece with Kevlar™ head harness, Quick disconnect, EZ-Flo Plus CBRN Regulator, Soft Case, HUD, RIC/UAC.
Description
Description
  • Time-tested, fire-proven Scott Air-Pak™ SCBA components
  • Unique all black design
  • Redundant pressure reducer with automatic back-up
  • Tactile feel end of service indicator (EOSI)
  • Black hardware and cylinder bag
  • Lightweight cylinder options in 2216 and 4500 psi, 30, 45 and 60 minute durations
  • Interface with all Scott communications
Specifications
Specifications
NIOSH
Swat-Pak™ SCBA
Swat-pak™ scba, av-2000™ comfort seal facepiece with kevlar head harness, quick-disconnect, e-z flo™ plus cbrn regulator, soft case, hud, ric/uac
4500psi
Designed for optimum protection during high risk search and rescue operation, hazardous chemicals or biological environments
Kevlar