Learn More
Scott Safety™ Swat-Pak™ SCBA First Responder Edition Available on GSA/VA Contract for Federal Government customers only.
Designed for optimum protection protection during high risk search and rescue operations, hazardous chemicals or biological environments
Manufacturer:  Scott Safety™ 805454418
 View more versions of this product
Catalog No. 19-121-446
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Swat-Pak SCBA, AV-2000 Comfort Seal facepiece with Kevlar™ head harness, Quick disconnect, EZ-Flo Plus CBRN Regulator, Soft Case, HUD, RIC/UAC.
Description
Description
  • Time-tested, fire-proven Scott Air-Pak™ SCBA components
  • Unique all black design
  • Redundant pressure reducer with automatic back-up
  • Tactile feel end of service indicator (EOSI)
  • Black hardware and cylinder bag
  • Lightweight cylinder options in 2216 and 4500 psi, 30, 45 and 60 minute durations
  • Interface with all Scott communications
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
NIOSH
Swat-Pak™ SCBA
Swat-pak™ scba, av-2000™ comfort seal facepiece with kevlar head harness, quick-disconnect, e-z flo™ plus cbrn regulator, soft case, hud, ric/uac
4500psi
Designed for optimum protection during high risk search and rescue operation, hazardous chemicals or biological environments
Kevlar