Learn More
Scott Safety™ Xcel™ Half-Mask Respirator
Ideal for almost any industrial, agricultural, or manufacturing application
Supplier: Scott Safety™ 7421113
Requires : Accepts any of the Scott 742 Series of low-profile, high-performance twin cartridges and filters (sold separately) to protect against gases, vapors, fumes, and particulates—separately and in combination.
Description
- Molded facepiece of thermoplastic elastomer (TPE) material combines soft, flexible comfort with durability and features a wide-face seal surface
- Unique “fold” over the bridge of the nose acts as shock absorber for normal facial movement such as talking, keeping the mask in place and the seal secure
- A choice of four sizes and an easily adjustable cradle suspension ensure the wearer enjoys an excellent fit and superior wearability
- Twin cartridges are mounted out of the line of vision so the wearer's view is not obstructed
- Head harness uses a convenient one-step adjustment procedure that can be done even when wearing gloves
- Will not interfere with glasses, goggles, headgear, or ear protection
- Optional Voicemitter allows clear communication; technically advanced transmitter improves worker communication and efficiency by providing outstanding voice clarity
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters; for use in atmospheres that are not Immediately Dangerous to Life and Health (non-IDLH) environments.
Specifications
NIOSH 42 CFR Part 84 | |
Half-Mask Respirator | |
Industrial, agricultural or manufacturing applications | |
Xcel facepiece assembly | |
Yolk and cradle |
Thermoplastic Elastomer | |
Medium/Large | |
Cartridge in atmospheres that are not immediately dangerous to life and health environments | |
7421-113 |