Learn More
Scott Safety™ Xcel™ Half-Mask Respirator
Ideal for almost any industrial, agricultural, or manufacturing application
Manufacturer:  Scott Safety™ 7421112V
 View more versions of this product
Catalog No. 19-700-833
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Requires : Accepts any of the Scott 742 Series of low-profile, high-performance twin cartridges and filters (sold separately) to protect against gases, vapors, fumes, and particulates—separately and in combination.
Description
Description
  • Molded facepiece of thermoplastic elastomer (TPE) material combines soft, flexible comfort with durability and features a wide-face seal surface
  • Unique “fold” over the bridge of the nose acts as shock absorber for normal facial movement such as talking, keeping the mask in place and the seal secure
  • A choice of four sizes and an easily adjustable cradle suspension ensure the wearer enjoys an excellent fit and superior wearability
  • Twin cartridges are mounted out of the line of vision so the wearer's view is not obstructed
  • Head harness uses a convenient one-step adjustment procedure that can be done even when wearing gloves
  • Will not interfere with glasses, goggles, headgear, or ear protection
  • Optional Voicemitter allows clear communication; technically advanced transmitter improves worker communication and efficiency by providing outstanding voice clarity
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters; for use in atmospheres that are not Immediately Dangerous to Life and Health (non-IDLH) environments.
Specifications
Specifications
Yolk and cradle
Safety
NIOSH 42CFR8
Cartridge in atmospheres that are not immediately dangerous to life and health environments
Thermoplastic Elastomer, Elastic
Half Mask Respirator
Small/Medium
Ideal for almost any industrial, agricultural, or manufacturing application
Xcel facepiece assembly, speaking diaphragm, voicemitter