Learn More
Scott Safety™ Xcel™ Half-Mask Respirator
Ideal for almost any industrial, agricultural, or manufacturing application
Manufacturer:  Scott Safety™ 7421114V
 View more versions of this product
Catalog No. 19-700-837
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Requires : Accepts any of the Scott 742 Series of low-profile, high-performance twin cartridges and filters (sold separately) to protect against gases, vapors, fumes, and particulates—separately and in combination.
Description
Description
  • Molded facepiece of thermoplastic elastomer (TPE) material combines soft, flexible comfort with durability and features a wide-face seal surface
  • Unique “fold” over the bridge of the nose acts as shock absorber for normal facial movement such as talking, keeping the mask in place and the seal secure
  • A choice of four sizes and an easily adjustable cradle suspension ensure the wearer enjoys an excellent fit and superior wearability
  • Twin cartridges are mounted out of the line of vision so the wearer's view is not obstructed
  • Head harness uses a convenient one-step adjustment procedure that can be done even when wearing gloves
  • Will not interfere with glasses, goggles, headgear, or ear protection
  • Optional Voicemitter allows clear communication; technically advanced transmitter improves worker communication and efficiency by providing outstanding voice clarity
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters; for use in atmospheres that are not Immediately Dangerous to Life and Health (non-IDLH) environments.
Specifications
Specifications
Yolk and cradle
Safety
NIOSH 42CFR8
Cartridge in atmospheres that are not immediately dangerous to life and health environments
Thermoplastic Elastomer, Elastic
Half Mask Respirator
Large
Ideal for almost any industrial, agricultural, or manufacturing application
Xcel facepiece assembly, speaking diaphragm, voicemitter