Learn More
Scott Safety™ Xcel™ Half-Mask Respirator
Ideal for almost any industrial, agricultural, or manufacturing application
Specifications
Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
---|---|
Material | Thermoplastic Elastomer |
Product Type | Half-Mask Respirator |
For Use With (Application) | Industrial, agricultural or manufacturing applications |
For Use With (Equipment) | Cartridge in atmospheres that are not immediately dangerous to life and health environments |
Requires: Accepts any of the Scott 742 Series of low-profile, high-performance twin cartridges and filters (sold separately) to protect against gases, vapors, fumes, and particulates—separately and in combination.
Catalog Number | Mfr. No. | Size | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Includes | Price | Quantity & Availability | ||||
19-700-823
|
Scott Safety™
7421111 |
Small | Xcel facepiece assembly | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
19-700-825
|
Scott Safety™
7421112 |
Small/Medium | Xcel facepiece assembly | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
19-700-827
|
Scott Safety™
7421113 |
Medium/Large | Xcel facepiece assembly | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
19-700-829
|
Scott Safety™
7421114 |
Large | Xcel facepiece assembly | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
19-700-831
|
Scott Safety™
7421111V |
Small | Xcel facepiece assembly, voicemitter | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
19-700-833
|
Scott Safety™
7421112V |
Small/Medium | Xcel facepiece assembly, speaking diaphragm, voicemitter | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
19-700-835
|
Scott Safety™
7421113V |
Medium/Large | Xcel facepiece assembly, speaking diaphragm, voicemitter | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
19-700-837
|
Scott Safety™
7421114V |
Large | Xcel facepiece assembly, speaking diaphragm, voicemitter | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
Description
- Molded facepiece of thermoplastic elastomer (TPE) material combines soft, flexible comfort with durability and features a wide-face seal surface
- Unique “fold” over the bridge of the nose acts as shock absorber for normal facial movement such as talking, keeping the mask in place and the seal secure
- A choice of four sizes and an easily adjustable cradle suspension ensure the wearer enjoys an excellent fit and superior wearability
- Twin cartridges are mounted out of the line of vision so the wearer's view is not obstructed
- Head harness uses a convenient one-step adjustment procedure that can be done even when wearing gloves
- Will not interfere with glasses, goggles, headgear, or ear protection
- Optional Voicemitter allows clear communication; technically advanced transmitter improves worker communication and efficiency by providing outstanding voice clarity
Certifications
NIOSH approved under 42CFR84 for use against a variety of respiratory hazards, when used as designed with appropriate cartridges and filters; for use in atmospheres that are not Immediately Dangerous to Life and Health (non-IDLH) environments.
Specifications
NIOSH 42 CFR Part 84 | |
Half-Mask Respirator | |
Cartridge in atmospheres that are not immediately dangerous to life and health environments |
Thermoplastic Elastomer | |
Industrial, agricultural or manufacturing applications | |
Yolk and cradle |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.