Learn More
Scott Safety™ Xcel™ Half-Mask Respirator
Ideal for almost any industrial, agricultural, or manufacturing application
Manufacturer:  Scott Health & Safety 7421111
 View more versions of this product
Catalog No. 19-700-823
  Check Availability
Add to cart
Requires : Accepts any of the Scott 742 Series of low-profile, high-performance twin cartridges and filters (sold separately) to protect against gases, vapors, fumes, and particulates—separately and in combination.
Specifications Description & Specifications
Specifications
NIOSH 42CFR8
Cartridge in atmospheres that are not immediately dangerous to life and health environments
Thermoplastic Elastomer, Elastic
Half Mask Respirator
Ideal for almost any industrial, agricultural, or manufacturing application
Xcel facepiece assembly
Yolk and cradle
Small