Scott Safety™ Xcel™ Half-Mask Respirator
Ideal for almost any industrial, agricultural, or manufacturing application
Manufacturer: Scott Health & Safety 7421113V
 View more versions of this product
Catalog No. 19-700-835
  Check Availability
Add to Cart
Required Accessories : Accepts any of the Scott 742 Series of low-profile, high-performance twin cartridges and filters (sold separately) to protect against gases, vapors, fumes, and particulates—separately and in combination.
    Specifications
    Certifications NIOSH 42CFR8
    For Use With (Application) Ideal for almost any industrial, agricultural, or manufacturing application
    For Use With (Equipment) Cartridge in atmospheres that are not immediately dangerous to life and health environments
    Includes Xcel facepiece assembly, speaking diaphragm, voicemitter
    Material Thermoplastic Elastomer, Elastic
    Suspension Type Yolk and cradle
    Product Type Half Mask Respirator
    Size Medium/Large
  • Description & Specifications
Related Products
Product Content Feedback