Learn More
SCP3/SYCP3, Mouse anti-Human, HRP, Clone: 6F9C5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP223487H100UL
Catalog No. NB005138
Add to cart
Description
Description
SCP3/SYCP3 Monoclonal specifically detects SCP3/SYCP3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
SCP3/SYCP3
6F9C5
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
SYCP3
Partial recombinant human SCP3 (between residues 1-200) expressed in E. coli [Uniprot: Q8IZU3]
Ammonium sulfate precipitation
RUO
Store at 4°C in the dark.
Monoclonal
Human
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
HRP
PBS
MGC71888, SCP3, SCP-3, SCP3 chromosome 3 open reading frame 8, SCP3 COR1, synaptonemal complex protein 3
Mouse
IgG1
100 uL
Cell Biology, Cell Cycle and Replication, Chromatin Research, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
50511