Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Screw Cap for Leucosep™ Tubes

For use with Leucosep, Polystyrene and Polypropylene tubes

$762.50

Specifications

Type Screw Cap
For Use With (Equipment) Leucosep, Polystyrene and Polypropylene Tubes
View More Specs

Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
07-000-514
View Documents
Greiner Bio-One
315376
White
Case of 9000 for $762.50
Add to cart
 
Description

Description

  • For use with Leucosep, Polystyrene and Polypropylene tubes
  • Color: Red
Specifications

Specifications

Screw Cap
Leucosep, Polystyrene and Polypropylene Tubes
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit