Thermo Scientific™ Screw Vial Caps and Seals Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
Manufacturer: Thermo Scientific™ 60180-733
 View more versions of this product
Catalog No. 03-250-641
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Description Blue cap with ivory PTFE/red rubber seal; Varian, Waters, Spark
    Closure Color Blue
    Type Screw Vial Caps and Seals
    Septum Ivory PTFE/red rubber seal
    For Use With Varian Autosamplers 9090/9095/9100/PROSTAR, Waters 96/Alliance, 2690/Acquity, Spark Symbiosis/Reliance
    Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
  • Description & Specifications
Related Products
Product Content Feedback