Thermo Scientific™ Screw Vial Caps and Seals Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
Manufacturer: Thermo Scientific™ 60180738
 View more versions of this product
Catalog No. 03-251-319
 N/A
    N/A
Part Not Found 
    Specifications
    Description Black cap with red PTFE/white silicone seal; PE, Claris, Shimadzu, Varian, Waters instruments
    Closure Color Black
    Septum red PTFE/white silicone seal
    Type Screw Vial Caps and Seals
    For Use With PE Autosystem/AS-2000, Claris 500, Shimadzu SIL-2A, SIL-10A, SIL-HT,/104DVP, Varian Autosamplers 8000/8100/8410, Waters 48/717/717 Plus
    Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
  • Description & Specifications
Related Products
Product Content Feedback