Learn More
Thermo Scientific™ Screw Vial Caps and Seals
Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
Manufacturer:  Thermo Scientific™ 60180718
 View more versions of this product
Catalog No. 03-250-626
  Check Availability
Add to cart
Specifications
Description
Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
Description & Specifications
Specifications
Thermo Scientific Spectra System AS3500/AS3000/AS1000/Surveyor/Accela LC, Agilent 1090, PE Autosystem/AS-2000, Claris 500, ISS-100, ISS-200, Integral 4000 & Series 200, Shimadzu AOC-20, AOC-14/1400, AOC-8B/9, AOC5000, SIL-2A, SIL-10A, SIL-HT,/104DVP, Varian Autosamplers 8000/8100/8410, 9090/9095/9100/PROSTAR, Waters 96/Alliance, 2690/Acquity, HItachi AS-1000/AS-2000/AS-4000
Screw Vial Seals
Red PTFE/white silicone seal; Thermo Scientific, Agilent, Claris, Shimadzu, Varian, Waters, Hitachi
Red PTFE/white silicone seal