Learn More
Tocris Bioscience™ Scriptaid
Histone deacetylase inhibitor
Manufacturer:  Tocris Bioscience™ 2421/50
 View more versions of this product
Catalog No. 24-215-0
Add to cart
This item is not returnable. View return policy
Description
Description
Novel histone deacetylase inhibitor. Produces > 100-fold increase in histone acetylation and displays relatively low toxicity.
Specifications
Specifications
Scriptaid
>99
50mg
Scriptaid
326.35