Learn More
Labconco™ ScrubAir™ Serological Pipette Insert for ScrubAir Pipette Washer/Dryers
Easily adapt your ScrubAir™ Pipette Washer/Dryer to permit cleaning of serological pipettes with the Labconco™ Serological Pipette Insert.
Manufacturer:  Labconco™ 4611900
Catalog No. 10-359-260
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Insert holds standard serological pipettes up to 50 mL, allowing them to be washed in the ScrubAir™
  • Holds up to 36 pipettes
  • Installs tool-free in seconds
Specifications
Specifications
36 pipettes
2.75 in.
6.5 in. Diameter
ScrubAir™ Pipette Washer/Dryer
Insert
1.5 lb