Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SCTR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA551056
Description
SCTR Polyclonal Antibody detects endogenous levels of SCTR protein.
SCTR Polyclonal antibody specifically detects SCTR in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
SCTR | |
Polyclonal | |
Unconjugated | |
Sctr | |
6530402O03Rik; MYOP; pancreatic secretin receptor; Sctr; SCT-R; Secretin receptor; SR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6344 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P47872 | |
Sctr | |
Synthesized peptide derived from the Internal region of human Secretin Receptor. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction