Learn More
SCYL2 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304880A

Description
The recommended shelf life for this product is 1 year from date of receipt.
SCYL2 is a component of AP2-containing clathrin coated structures at the plasma membrane or of endocytic coated vesicles. SCYL2 is a probable serine/threonine-protein kinase that phosphorylates, in vitro, the beta2-subunit of the plasma membrane adapter complex AP2 and other proteins in presence of poly-L-lysine. SCYL2 may also regulate clathrin-dependent trafficking between the TGN and/or the endosomal system.Specifications
SCYL2 | |
Polyclonal | |
Unconjugated | |
SCYL2 | |
SCY1-like protein 2; Coated vesicle-associated kinase of 104 kDa; SCYL2; CVAK104; KIAA1360; | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
213326, 55681 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q6P3W7, Q8CFE4 | |
SCYL2 | |
Between 850 and 900 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.