Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SCYL3 Monoclonal Antibody (OTI4F10)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA524934

Catalog No. PIMA524934


Only null left
Add to Cart

Description

Description

SCYL3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

May play a role in regulating cell adhesion/migration complexes in migrating cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SCYL3
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8IZE3
SCYL3
Full-length protein expressed in 293T cell transfected with human SCYL3 expression vector.
100 μL
Primary
Human, Monkey
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI4F10
Unconjugated
SCYL3
1200016D23Rik; 6030457O16; AW214499; ezrin-binding partner PACE-1; ezrin-binding partner PACE-1 (PACE-1); ezrin-binding protein PACE-1; Pace1; PACE-1; protein-associating with the carboxyl-terminal domain of ezrin; RP1-97P20.2; SCY1 like pseudokinase 3; SCY1-like 3; SCY1-like 3 (S. cerevisiae); SCY1-like protein 3; SCY1-like, kinase-like 3; Scyl3
Mouse
Affinity chromatography
RUO
57147
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.