Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDAD1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549391

Catalog No. PA549391

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, zebrafish and rat based on sequence homology.

Required for 60S pre-ribosomal subunits export to the cytoplasm.
Specifications

Specifications

SDAD1
Polyclonal
PBS with 0.09% sodium azide
Q9NVU7, Q80UZ2
Protein SDA1 homolog, Nucleolar protein 130, SDA1 domain-containing protein 1, hSDA, SDAD1, NUC130
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
231452, 55153
Western Blot
Unconjugated
SDAD1
Liquid
SDAD1
KLH conjugated synthetic peptide between 596-629 amino acids from the C-terminal region of human SDAD1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit