Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDCBP Mouse anti-Human, Clone: OTI2H6, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA504796
Description
The protein encoded by this gene was initially identified as a molecule linking syndecan-mediated signaling to the cytoskeleton. The syntenin protein contains tandemly repeated PDZ domains that bind the cytoplasmic, C-terminal domains of a variety of transmembrane proteins. This protein may also affect cytoskeletal-membrane organization, cell adhesion, protein trafficking, and the activation of transcription factors. The protein is primarily localized to membrane-associated adherens junctions and focal adhesions but is also found at the endoplasmic reticulum and nucleus. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
SDCBP | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
O00560 | |
MDA-9, MDA9, ST1, SYCL, TACIP18 | |
Mouse | |
IgG2b | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
6386 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI2H6 | |
Unconjugated | |
SDCBP | |
Liquid | |
SDCBP | |
Full length human recombit protein of human SDCBP produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title