Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDCCAG8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551296

Catalog No. PA551296


Add to cart

Description

Description

The antibody detects endogenous levels of total SDCCAG8 protein.

This gene encodes a centrosome associated protein. This protein may be involved in organizing the centrosome during interphase and mitosis. Mutations in this gene are associated with retinal-renal ciliopathy.
Specifications

Specifications

SDCCAG8
Polyclonal
Unconjugated
SDCCAG8
Liquid
SDCCAG8
Synthetic peptide of human SDCCAG8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
0.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q86SQ7
CCCAP, NPHP10
Rabbit
IgG
100 μL
-20°C
Primary
10806
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit