Learn More
Gibco™ SDF1B Recombinant Human Protein
$368.00
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
PHC1364 Non-distribution product as customer accommodation.
View Documents
Gibco™
PHC1364
N/A 10ug
Each for $368.00
Add to cart
Description
Description
SDF1B is a small cytokine that belongs to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1.