Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SDF4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552809
Description
Immunogen sequence: DQDGDKQLSV PEFISLPVGT VENQQGQDID DNWVKDRKKE FEELIDSNHD GIVTAEEPQS YMDLRRAPPL SNN Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 79%.
This gene encodes a stromal cell derived factor that is a member of the CREC protein family. The encoded protein contains six EF-hand motifs and calcium-binding motifs. This protein localizes to the Golgi lumen and may be involved in regulating calcium dependent cellular activities.
Specifications
SDF4 | |
Polyclonal | |
Unconjugated | |
SDF4 | |
45 kDa calcium-binding protein; cab45; PSEC0034; Sdf4; SDF-4; stromal cell derived factor 4; stromal cell-derived factor 4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRK5 | |
SDF4 | |
Recombinant protein corresponding to Human SDF4. Recombinant protein control fragment (Product #RP-89994). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction