Learn More
Invitrogen™ SDHA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579963
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human MCF-7 whole cell, human HepG2 whole cell, rat brain tissue, mouse brain tissue, mouse spleen tissue. IHC: human lung cancer tissue, human breast cancer tissue, human spleen tissue, human colorectal adenocarcinoma tissue, human rectum adenocarcinoma tissue. Flow: PC-3 cell.
This gene encodes a major catalytic subunit of succinate-ubiquinone oxidoreductase, a complex of the mitochondrial respiratory chain. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. Mutations in this gene have been associated with a form of mitochondrial respiratory chain deficiency known as Leigh Syndrome. A pseudogene has been identified on chromosome 3q29.
Specifications
SDHA | |
Polyclonal | |
Unconjugated | |
SDHA | |
1500032O14Rik; 2310034D06Rik; 4921513A11; C81073; CMD1GG; Flavoprotein subunit of complex II; FP; im:7141001; mitochondrial succinate dehydrogenase complex subunit A; PGL5; SDH1; SDH2; SDHA; SDHA protein; SDHF; SDHFP1; si:dkeyp-84f11.2; succinate dehydrogenase [ubiquinone] flavoprotein subunit; succinate dehydrogenase [ubiquinone] flavoprotein subunit, mitochondrial; succinate dehydrogenase complex flavoprotein subunit A; succinate dehydrogenase complex subunit A; succinate dehydrogenase complex subunit A flavoprotein; succinate dehydrogenase complex subunit A, flavoprotein (Fp); succinate dehydrogenase complex, subunit A, flavoprotein (Fp); succinate dehydrogenase flavoprotein subunit; succinate dehydrogenase flavoprotein subunit A; zgc:56051 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
157074, 6389, 66945 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
P31040, Q8K2B3, Q920L2 | |
SDHA | |
A synthetic peptide corresponding to a sequence at the N-terminus of human SDHA (113-128aa NMEEDNWRWHFYDTVK). | |
100 μg | |
Primary | |
Human, Mouse, Rat, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.