Learn More
SDHAF1 Mouse anti-Human, Clone: OTI8C12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809693
Catalog No. 50-166-9660
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
SDHAF1 (Succinate dehydrogenase complex assembly factor 1) is one of the subunits of the succinate dehydrogenase (SDH) complex (or complex II) of the mitochondrial respiratory chain. SDHAF1 resides in the mitochondria, and is essential for SDH assembly, but does not physically associate with the complex in vivo. Mutations in this gene are associated with SDH-defective infantile leukoencephalopathy (mitochondrial complex II deficiency). Unlike the related protein SDHAF2, SDHAF1 is not thought to be a tumor suppressor.
Specifications
Specifications
SDHAF1
Monoclonal
Unconjugated
SDHAF1
Lyophilized
SDHAF1
Full length human recombit protein of human SDHAF1 produced in HEK293T.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI8C12
PBS with 8% trehalose and no preservative; pH 7.3
A6NFY7
LYRM8
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
644096