Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDHB Mouse anti-Human, Clone: SDHB/2382, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen SDHB
Clone SDHB/2382
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Protein Array
Applications Western Blot, Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123047
missing translation for 'viewDocuments'
Novus Biologicals
NBP30778710UG
100 μg
Each for $352.89
Add to cart
 
NB123048
missing translation for 'viewDocuments'
Novus Biologicals
NBP30778720UG
20 μg
Each for $138.37
Add to cart
 
Description

Description

SDHB Monoclonal antibody specifically detects SDHB in Human samples. It is validated for Western Blot, Protein Array
Specifications

Specifications

SDHB
0.2 mg/ml
Western Blot, Peptide Array
Purified
RUO
Human
EC 1.3.5.1, FLJ92337, IP, Iron-sulfur subunit of complex II, PGL4, SDH, SDH1, SDH2, SDHIP, succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial, succinate dehydrogenase complex, subunit B, iron sulfur (Ip)
Recombinant fragment (around aa 165-273) of human SDHB protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
SDHB/2382
Western Blot 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Cancer
10 mM PBS with 0.05% BSA
6390
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit