Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SDR9C7 Monoclonal Antibody (OTI6A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Supplier: OriGene TA501706
Description
SDR9C7 encodes a protein with similarity to the short-chain dehydrogenase/reductase (SDR) family but has not been shown to have retinoid or dehydrogenase activities.Specifications
SDR9C7 | |
Monoclonal | |
0.8 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q8NEX9 | |
SDR9C7 | |
Full length human recombit protein of human SDR9C7 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
OTI6A8 | |
Unconjugated | |
SDR9C7 | |
ARCI13, RDHS, SDR-O, SDRO | |
Mouse | |
Affinity Chromatography | |
RUO | |
121214 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction