Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDR9C7 Mouse anti-Human, Clone: OTI8F3, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502013

Catalog No. 50-167-3101


Add to cart

Description

Description

SDR9C7 encodes a protein with similarity to the short-chain dehydrogenase/reductase (SDR) family but has not been shown to have retinoid or dehydrogenase activities.
Specifications

Specifications

SDR9C7
Monoclonal
0.74 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8NEX9
ARCI13, RDHS, SDR-O, SDRO
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
121214
Flow Cytometry, Western Blot
OTI8F3
Unconjugated
SDR9C7
Liquid
SDR9C7
Human recombit protein fragment corresponding to amino acids 1-313 of human produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit