Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZING™ Enterprises SDS Economy Station Kit

A variety of sizes and configurations are available.

Manufacturer:  ZING™ Enterprises 6042

 View more versions of this product

Catalog No. 18-003-007


Add to cart

Description

Description

ZING 2708 SDS Economy Station Kit includes
• Qty. 1 Clear Plastic Wall Rack
• Qty. 1 SDS Binder, recycled plastic, (1.5 in. Ring) holds up to 300 sheets
• Qty. 1 SDS Station Wall Sign, recycled plastic, measures 10 in.H x 14 in.W

ZING SDS Training Booklets support compliance with the new GHS/Hazard Communication requirements. Booklets are designed to support GHS training and awareness programs and can be used with other ZING SDS products to include binders and compliance stations. 10 per Pack. Made in the USA. Available in English or Spanish

ZING 6039 SDS Single Station contents include:
• Qty 1 SDS Station, recycled aluminum
• Qty 1 SDS Binder, recycled plastic, 1.5 in. ring, stores up to 300 sheets

ZING 6040 SDS Double Station contents include:
Qty 1 SDS Station, recycled aluminum
Qty 2 SDS Binder, recycled plastic, 1.5 in. ring, stores up to 300 sheets each

ZING 6041 SDS Training Station contents include:
• Qty 1 SDS Station, recycled aluminum
• Qty 1 SDS Binder, recycled plastic, 1.5 in. ring, stores up to 300 sheets
• Qty 10 training booklets (English)

ZING 6042 SDS Single (English/Spanish Bilingual) Station contents include:
• Qty 1 SDS Station, recycled aluminum
• Qty 1 Bilingual SDS Binder, recycled plastic, 1.5 in. ring, stores up to 300 sheets

ZING 6043 SDS Double (English/Spanish Bilingual) Station contents include:
• Qty 1 SDS Station, recycled aluminum, blingual
• Qty 2 SDS Binders, recycled plastic, 1.5’’ ring, stores up to 300 sheets, bilingual

ZING 6044 Training (English/Spanish Bilingual) Station contents include:
• Qty 1 SDS Station, bilingual, recycled aluminum
• Qty 1 SDS Binder, bilingual, recycled plastic, 1.5 in. ring, stores up to 300 sheets
• Qty 5 Training Booklets, English
• Qty 5 Training Booklets, Spanish

ZING 6066 SDS Training Center Station Contents Include:
• Qty. 1 - Binder with GHS Training Procedures (English)
• Qty. 1 - GHS Training DVD (English)
• Qty. 10 - Training Booklets (English)
• Qty. 1 - Right to Understand SDS Station, recycled aluminum

ZING 6067 SDS Training Center (Spanish) Station Contents Include:
• Qty. 1 - Right to Understand SDS Station, recycled aluminum (Bilingual)
• Qty. 1 - Binder with GHS Training Procedures (Spanish)
• Qty. 1 - GHS Training DVD (Spanish)
• Qty. 10 - Training Booklets (Spanish)

 

Specifications

Specifications

Red, White
Aluminum
20 in. x 5.25 in.
Corner Holes
20 in.
5.25 in.
English/Spanish
1
5.25 in.
SDS Management
Hazard Communication
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit