Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SDS Mouse anti-Human, Clone: OTI1C8, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503994

Catalog No. 50-167-4767


Add to cart

Description

Description

SDS encodes one of three enzymes that are involved in metabolizing serine and glycine. L-serine dehydratase converts L-serine to pyruvate and ammonia and requires pyridoxal phosphate as a cofactor. The encoded protein can also metabolize threonine to NH4+ and 2-ketobutyrate. The encoded protein is found predomitly in the liver.
Specifications

Specifications

SDS
Monoclonal
0.53 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P20132
SDH
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10993
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1C8
Unconjugated
SDS
Liquid
SDS
Full length human recombit protein of human SDS produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit