Learn More
SDZ 220-040, Tocris
Potent, competitive NMDA antagonist
Manufacturer:  R&D Systems 1251/10
Quantity 10mg
 View more versions of this product
Catalog No. 12-511-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
420.19
SDZ 220-040
C16H16Cl2NO6P
(S)-α-Amino-2', 4'-dichloro-4-hydroxy-5-(phosphonomethyl)-[1, 1'-biphenyl]-3-propanoic acid
Specifications
174575-40-7
>98%
420.19
SDZ 220-040
10mg
C16H16Cl2NO6P
(S)-α-Amino-2', 4'-dichloro-4-hydroxy-5-(phosphonomethyl)-[1, 1'-biphenyl]-3-propanoic acid
Safety and Handling
Recommended Storage : Store at Room Temperature