Learn More
Julabo USA Inc Each, SE-12 heating circulator +20…+300 °C, 3kW heater
SDP
Supplier:  Julabo USA Inc 9252512.33
Encompass_Preferred
Bath/Circulators, Julabo, SE-12 Heating Circulator +20...+300 °C; 12L bath tank with 3kW heater at 230V; 208-230V/50-60Hz; integrated cooling coil. Suitable for temperature control of internal and external applications.4-speed centrifugal pressure/suction pump providing 22-26 L/min with pressures from 5.8-10.2 psi / suction from 2.9-5.8 psi. M16x1 male fittings with 8 and 12 mm hose barbs; ±0.01 °C temperature stability. RS232 interface for PC integrationLarge TFT screen, Pt100 external sensor outlet for external measurement and control and an integrated programmer for 1x10 (HE/SE) or 6x60 (HL/SL) program steps. Includes port for optional analog interface (8900100).
Catalog No. 50-753-3939