Learn More
Medchemexpress LLC SE-7552 | 2243575-79-1 | 99.5% | 335.28 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY16130510MM/1ML
Encompass_Preferred
SE-7552, a 2-(difluoromethyl)-1,3,4-oxadiazole (DFMO) derivative, is an orally active, highly selective, non-hydroxamate HDAC6 inhibitor with an IC50 of 33 nM. It demonstrates greater than 850-fold selectivity versus all other known HDAC isozymes. This compound is capable of blocking multiple myeloma growth in vivo and acts as an anti-obesity agent in diet-induced obese mice.
  • High selectivity for HDAC6
  • Oral activity
  • Non-hydroxamate nature
  • Shown efficacy in blocking multiple myeloma growth in vivo
  • Acts as an anti-obesity agent
Catalog No. 50-003-91725